siRNA Details
| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142792.1 | NP_001136264.1 | 2 | 223 | ||
| NM_177887.4 | NP_808555.1 | 2 | 392 | |||
| GenBank mRNA | AK041846.1 | - | 2 | 393 | ||
| BC022651.1 | - | 2 | 223 | 4216812 | ||

Tmem150b
Genomic Map