siRNA Details
Gene Symbol: Teddm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178244.3 | NP_839975.2 | 1 | 592 | ||
GenBank mRNA | AJ554212.1 | - | 1 | 446 | ||
BC109332.2 | - | 1 | 445 | |||
BC141344.1 | - | 1 | 495 | 9055956 | ||
BC141345.1 | - | 1 | 496 | 9055957 |
Gene Symbol: Teddm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008426.3 | NP_001008426.1 | 1 | 551 | ||
GenBank mRNA | AK033834.1 | - | 1 | 552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
