siRNA Details
| Gene Symbol: Rnf152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160368.1 | NP_001153840.1 | 2 | 1896 | ||
| NM_178779.4 | NP_848894.1 | 3 | 2019 | |||
| GenBank mRNA | AK044638.1 | - | 3 | 1978 | ||
| AK081452.1 | - | 1 | 47 | |||
| BC118961.2 | - | 2 | 1640 | 40094497 | ||

Rnf152
Genomic Map