siRNA Details
| Gene Symbol: RAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308353.1 | NP_001295282.1 | 4 | 803 | ||
| NM_005855.3 | NP_005846.1 | 3 | 742 | |||
| XM_017003152.1 | XP_016858641.1 | 3 | 664 | |||
| XM_017003153.1 | XP_016858642.1 | 3 | 972 | |||
| XM_017003154.1 | XP_016858643.1 | 3 | 749 | |||
| XM_017003156.1 | XP_016858645.1 | 3 | 690 | |||
| GenBank mRNA | AJ001014.1 | - | 3 | 642 | ||
| BC000548.2 | - | 3 | 634 | 3163522 | ||
| BI548986.1 | - | 4 | 728 | |||

RAMP1
Genomic Map