siRNA Details
| Gene Symbol: Sdr16c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181989.1 | NP_871789.1 | 7 | 1093 | ||
| GenBank mRNA | AK136508.1 | - | 6 | 1064 | ||
| AY277588.1 | - | 7 | 1093 | |||
| BC104134.1 | - | 7 | 1012 | 40045268 | ||
| BC104135.1 | - | 7 | 1012 | 40045269 | ||

Sdr16c5
Genomic Map