siRNA Details
| Gene Symbol: Tmem150c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182841.1 | NP_878261.1 | 8 | 1082 | ||
| GenBank mRNA | AK044570.1 | - | 8 | 843 | ||
| BC028943.1 | - | 3 | 403 | |||
| BC055321.1 | - | 8 | 1082 | 6419443 | ||
| BC062109.1 | - | 8 | 860 | |||

Tmem150c
Genomic Map