siRNA Details
| Gene Symbol: Tmem150c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182841.1 | NP_878261.1 | 8 | 2783 | ||
| GenBank mRNA | BC028943.1 | - | 3 | 2104 | ||
| BC055321.1 | - | 8 | 2783 | 6419443 | ||
| BC062109.1 | - | 8 | 2561 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem150c| Gene Symbol: Tmem150c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182841.1 | NP_878261.1 | 8 | 2783 | ||
| GenBank mRNA | BC028943.1 | - | 3 | 2104 | ||
| BC055321.1 | - | 8 | 2783 | 6419443 | ||
| BC062109.1 | - | 8 | 2561 | |||