siRNA Details
| Gene Symbol: Atxn2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183020.1 | NP_898841.1 | 15 | 2119 | ||
| GenBank mRNA | AK155062.1 | - | 15 | 2166 | ||
| AK158626.1 | - | 15 | 1806 | |||
| AK168745.1 | - | 15 | 1836 | |||
| BC034083.1 | - | 9 | 1198 | |||
| BC043451.1 | - | 10 | 1277 | |||
| BC054483.1 | - | 15 | 2119 | 6391591 | ||

Atxn2l
Genomic Map