siRNA Details
Gene Symbol: A430033K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183025.2 | NP_898846.2 | 4 | 975 | ||
GenBank mRNA | AK039936.1 | - | 961 | |||
BC055299.1 | - | 4 | 940 | 6492709 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A430033K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183025.2 | NP_898846.2 | 4 | 975 | ||
GenBank mRNA | AK039936.1 | - | 961 | |||
BC055299.1 | - | 4 | 940 | 6492709 |