siRNA Details
| Gene Symbol: Rnase9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183032.2 | NP_898853.2 | 2 | 292 | ||
| GenBank mRNA | AK078986.1 | - | 2 | 295 | ||
| AY226989.1 | - | 2 | 293 | |||
| AY303816.1 | - | 2 | 292 | |||
| BC107218.1 | - | 2 | 297 | 40051762 | ||

Rnase9
Genomic Map