siRNA Details
| Gene Symbol: Elfn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183141.2 | NP_898964.2 | 2 | 1221 | ||
| GenBank mRNA | AK031970.1 | - | 1 | 811 | ||
| BC076582.1 | - | 2 | 1462 | |||
| BC079588.1 | - | 1 | 721 | 5706857 | ||
| BC094219.1 | - | 2 | 1221 | 6840886 | ||

Elfn2
Genomic Map