siRNA Details
| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048204.1 | NP_001041669.1 | 4 | 2137 | ||
| GenBank mRNA | AK028336.1 | - | 3 | 1982 | ||
| BC010651.1 | - | 1 | 1584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp455| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048204.1 | NP_001041669.1 | 4 | 2137 | ||
| GenBank mRNA | AK028336.1 | - | 3 | 1982 | ||
| BC010651.1 | - | 1 | 1584 | |||