siRNA Details
| Gene Symbol: Cyp2ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183158.3 | NP_898981.2 | 8 | 1065 | ||
| GenBank mRNA | AK089408.1 | - | 8 | 1068 | ||
| BC138088.1 | - | 4 | 584 | 8861107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2ab1| Gene Symbol: Cyp2ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183158.3 | NP_898981.2 | 8 | 1065 | ||
| GenBank mRNA | AK089408.1 | - | 8 | 1068 | ||
| BC138088.1 | - | 4 | 584 | 8861107 | ||