siRNA Details
| Gene Symbol: Gm4884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183166.2 | NP_898989.2 | 3 | 2591 | ||
| GenBank mRNA | AK029779.1 | - | 3 | 2592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4884| Gene Symbol: Gm4884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183166.2 | NP_898989.2 | 3 | 2591 | ||
| GenBank mRNA | AK029779.1 | - | 3 | 2592 | ||