siRNA Details
| Gene Symbol: Homez | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177705.1 | NP_001171176.1 | 2 | 1315 | ||
| NM_183174.3 | NP_898997.2 | 2 | 1160 | |||
| GenBank mRNA | AK129361.1 | - | 2 | 1205 | ||
| AY258064.1 | - | 2 | 1263 | |||
| BC094669.1 | - | 2 | 1160 | 30616315 | ||

Homez
Genomic Map