siRNA Details
| Gene Symbol: Zfp300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183185.3 | NP_899008.2 | 4 | 449 | ||
| GenBank mRNA | AK053018.1 | - | 4 | 450 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp300| Gene Symbol: Zfp300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183185.3 | NP_899008.2 | 4 | 449 | ||
| GenBank mRNA | AK053018.1 | - | 4 | 450 | ||