siRNA Details
| Gene Symbol: Slfn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183201.4 | NP_899024.3 | 3 | 622 | ||
| GenBank mRNA | AK036486.1 | - | 3 | 623 | ||
| AK039815.1 | - | 3 | 611 | |||
| AK040359.1 | - | 3 | 609 | |||
| AK171886.1 | - | 2 | 564 | |||
| AY261804.1 | - | 2 | 584 | |||
| AY261805.1 | - | 2 | 579 | |||
| BC117909.1 | - | 2 | 502 | 40097236 | ||
| BC117910.1 | - | 2 | 502 | 40097237 | ||

Slfn5
Genomic Map