siRNA Details
| Gene Symbol: Zmiz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183208.3 | NP_899031.2 | 11 | 1837 | ||
| GenBank mRNA | AK054366.1 | - | 11 | 1730 | ||
| BC057691.1 | - | 1 | 74 | |||
| BC058646.1 | - | 11 | 1737 | 6405777 | ||
| BC065120.1 | - | 11 | 1730 | 6856060 | ||

Zmiz1
Genomic Map