siRNA Details
| Gene Symbol: BC107364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085699.1 | - | 4 | 643 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC107364| Gene Symbol: BC107364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085699.1 | - | 4 | 643 | ||