siRNA Details
| Gene Symbol: BC107364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256180.1 | NP_001243109.1 | 5 | 931 | ||
| NM_001256181.1 | NP_001243110.1 | 4 | 837 | |||
| GenBank mRNA | AK085699.1 | - | 6 | 1250 | ||
| BC107364.1 | - | 4 | 837 | 40056165 | ||
| BC107365.1 | - | 5 | 931 | 40056172 | ||

BC107364
Genomic Map