siRNA Details
| Gene Symbol: Gm5115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086292.1 | - | 1133 | |||
| BC145760.1 | - | 1 | 698 | 40131063 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5115| Gene Symbol: Gm5115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086292.1 | - | 1133 | |||
| BC145760.1 | - | 1 | 698 | 40131063 | ||