siRNA Details
| Gene Symbol: Btbd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081038.2 | NP_001074507.2 | 5 | 684 | ||
| GenBank mRNA | AK087912.1 | - | 5 | 686 | ||
| BB559682.2 | - | 3 | 243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd16| Gene Symbol: Btbd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081038.2 | NP_001074507.2 | 5 | 684 | ||
| GenBank mRNA | AK087912.1 | - | 5 | 686 | ||
| BB559682.2 | - | 3 | 243 | |||