siRNA Details
Gene Symbol: Khdc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC141037.1 | - | 999 | 9055649 |
Gene Symbol: Khdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183322.2 | NP_899145.2 | 3 | 1316 | ||
GenBank mRNA | AK162365.1 | - | 1 | 412 | ||
AY238603.1 | - | 3 | 1079 |
