siRNA Details
| Gene Symbol: Cachd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198037.1 | NP_932154.1 | 1, 2 | 700 | ||
| GenBank mRNA | AK046646.1 | - | 616 | |||
| AK129397.1 | - | 1, 2 | 73 | |||
| BC058676.1 | - | 1, 2 | 700 | 6834428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cachd1| Gene Symbol: Cachd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198037.1 | NP_932154.1 | 1, 2 | 700 | ||
| GenBank mRNA | AK046646.1 | - | 616 | |||
| AK129397.1 | - | 1, 2 | 73 | |||
| BC058676.1 | - | 1, 2 | 700 | 6834428 | ||