siRNA Details
| Gene Symbol: Cachd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198037.1 | NP_932154.1 | 2, 3 | 761 | ||
| GenBank mRNA | AK046646.1 | - | 677 | |||
| AK080923.1 | - | 2, 3 | 766 | |||
| AK129397.1 | - | 2, 3 | 134 | |||
| BC058676.1 | - | 2, 3 | 761 | 6834428 | ||

Cachd1
Genomic Map