siRNA Details
| Gene Symbol: Rnf133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198251.2 | NP_937894.1 | 1 | 632 | ||
| GenBank mRNA | AK028082.1 | - | 1 | 633 | ||
| AY155441.1 | - | 1 | 261 | |||
| BC116423.1 | - | 1 | 533 | 40103909 | ||
| BC116424.1 | - | 1 | 533 | 40103912 | ||

Rnf133
Genomic Map