siRNA Details
| Gene Symbol: Ska3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198605.3 | NP_941007.1 | 3 | 369 | ||
| GenBank mRNA | AK089223.1 | - | 3 | 370 | ||
| AK158142.1 | - | 3 | 391 | |||
| AK166171.1 | - | 3 | 360 | |||
| BC117500.1 | - | 3 | 361 | 40105921 | ||
| BC117501.1 | - | 3 | 361 | 40105920 | ||

Ska3
Genomic Map