siRNA Details
| Gene Symbol: Gm5124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032684.1 | - | 2 | 1871 | ||
| Gene Symbol: Nolc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039351.2 | NP_001034440.1 | 13 | 3557 | ||
| NM_001039352.2 | NP_001034441.1 | 13 | 3563 | |||
| NM_001039353.2 | NP_001034442.1 | 13 | 3560 | |||
| NM_053086.3 | NP_444316.2 | 13 | 3560 | |||
| GenBank mRNA | AK034817.1 | - | 13 | 3437 | ||
| AK147118.1 | - | 13 | 3433 | |||

Gm5124
Genomic Map