siRNA Details
| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123370.1 | NP_001116842.1 | 2 | 191 | ||
| GenBank mRNA | AK042420.1 | - | 1 | 171 | ||
| AK078855.1 | - | 2 | 179 | |||
| AK160486.1 | - | 2 | 243 | |||
| AK161147.1 | - | 2 | 179 | |||
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039552.2 | NP_001034641.2 | 190 | |||
| GenBank mRNA | BB614944.1 | - | 191 | |||
| BC150877.1 | - | 251 | 9087788 | |||
| Gene Symbol: Gm3435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123372.1 | NP_001116844.1 | 587 | |||
| GenBank mRNA | AK013428.1 | - | 188 | |||
| AK078282.1 | - | 181 | ||||

9030025P20Rik
Genomic Map