siRNA Details
| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042420.1 | - | 7 | 1537 | ||
| AK161147.1 | - | 7 | 1399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030025P20Rik| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042420.1 | - | 7 | 1537 | ||
| AK161147.1 | - | 7 | 1399 | |||