siRNA Details
| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123370.1 | NP_001116842.1 | 4, 5 | 455 | ||
| GenBank mRNA | AK042420.1 | - | 3, 4 | 435 | ||
| AK078855.1 | - | 4, 5 | 443 | |||
| AK160486.1 | - | 4, 5 | 507 | |||
| AK161147.1 | - | 4, 5 | 443 | |||
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039552.2 | NP_001034641.2 | 454 | |||
| GenBank mRNA | BB614944.1 | - | 568 | |||
| BC150877.1 | - | 515 | 9087788 | |||
| Gene Symbol: Gm3435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123372.1 | NP_001116844.1 | 851 | |||
| GenBank mRNA | AK013428.1 | - | 1631 | |||
| AK078282.1 | - | 445 | ||||

9030025P20Rik
Genomic Map