siRNA Details
| Gene Symbol: CHST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166395.1 | NP_001159867.1 | 2 | 985 | ||
| NM_005769.2 | NP_005760.1 | 2 | 933 | |||
| GenBank mRNA | AF131235.1 | - | 2 | 903 | ||
| AF149783.1 | - | 1 | 814 | |||
| AF280088.1 | - | 2 | 843 | |||
| AK026635.1 | - | 2 | 793 | |||
| AK074746.1 | - | 2 | 790 | |||
| BC035282.1 | - | 2 | 799 | 5179826 | ||

CHST4
Genomic Map