siRNA Details
| Gene Symbol: Vmn2r-ps104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049221.1 | - | 2437 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r-ps104| Gene Symbol: Vmn2r-ps104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049221.1 | - | 2437 | |||