siRNA Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198967.5 | NP_945318.2 | 7, 8 | 1279 | ||
| GenBank mRNA | AK046396.1 | - | 7, 8 | 1280 | ||
| AK137545.1 | - | 7, 8 | 1251 | |||
| AK162239.1 | - | 7, 8 | 935 | |||
| BC145513.1 | - | 5, 6 | 610 | |||

Tmtc1
Genomic Map