siRNA Details
| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045559.1 | NP_001039024.1 | 4 | 724 | ||
| GenBank mRNA | BC051011.1 | - | 4 | 667 | ||
| BC057593.1 | - | 5 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp952| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045559.1 | NP_001039024.1 | 4 | 724 | ||
| GenBank mRNA | BC051011.1 | - | 4 | 667 | ||
| BC057593.1 | - | 5 | 834 | |||