siRNA Details
| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199025.2 | NP_950190.2 | 2 | 1897 | ||
| GenBank mRNA | AK044592.1 | - | 2 | 1528 | ||
| AK173205.1 | - | 2 | 1522 | |||
| BC060655.1 | - | 2 | 1897 | 30355035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb26| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199025.2 | NP_950190.2 | 2 | 1897 | ||
| GenBank mRNA | AK044592.1 | - | 2 | 1528 | ||
| AK173205.1 | - | 2 | 1522 | |||
| BC060655.1 | - | 2 | 1897 | 30355035 | ||