siRNA Details
| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 2 | 170 | ||
| GenBank mRNA | AK137244.1 | - | 3 | 397 | ||
| AK172988.1 | - | 2 | 81 | |||
| BC062951.1 | - | 2 | 171 | 6852711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep135| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 2 | 170 | ||
| GenBank mRNA | AK137244.1 | - | 3 | 397 | ||
| AK172988.1 | - | 2 | 81 | |||
| BC062951.1 | - | 2 | 171 | 6852711 | ||