siRNA Details
| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 4 | 560 | ||
| GenBank mRNA | AK137244.1 | - | 5 | 787 | ||
| AK172988.1 | - | 4 | 471 | |||
| BC062951.1 | - | 4 | 561 | 6852711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep135| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 4 | 560 | ||
| GenBank mRNA | AK137244.1 | - | 5 | 787 | ||
| AK172988.1 | - | 4 | 471 | |||
| BC062951.1 | - | 4 | 561 | 6852711 | ||