siRNA Details
| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 25 | 3846 | ||
| GenBank mRNA | BC062951.1 | - | 25 | 3847 | 6852711 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep135| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 25 | 3846 | ||
| GenBank mRNA | BC062951.1 | - | 25 | 3847 | 6852711 | |