siRNA Details
| Gene Symbol: Tsen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199033.1 | NP_950198.1 | 12 | 1639 | ||
| GenBank mRNA | AK134697.1 | - | 12 | 1635 | ||
| AK186269.1 | - | 1 | 40 | |||
| BC061473.1 | - | 12 | 1639 | 6414790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsen2| Gene Symbol: Tsen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199033.1 | NP_950198.1 | 12 | 1639 | ||
| GenBank mRNA | AK134697.1 | - | 12 | 1635 | ||
| AK186269.1 | - | 1 | 40 | |||
| BC061473.1 | - | 12 | 1639 | 6414790 | ||