siRNA Details
| Gene Symbol: Fbxw15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199036.2 | NP_950201.2 | 7, 8 | 1031 | ||
| GenBank mRNA | AK087669.1 | - | 7, 8 | 1032 | ||
| CO795020.1 | - | 1, 2 | 52 | |||
| DQ067445.1 | - | 7, 8 | 999 | |||
| Gene Symbol: Fbxw25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143305.1 | - | 7, 8 | 1029 | ||

Fbxw15
Genomic Map