siRNA Details
| Gene Symbol: Fbxw15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199036.2 | NP_950201.2 | 8 | 1047 | ||
| GenBank mRNA | AK087669.1 | - | 8 | 1048 | ||
| CO795020.1 | - | 2 | 68 | |||
| DQ067445.1 | - | 8 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw15| Gene Symbol: Fbxw15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199036.2 | NP_950201.2 | 8 | 1047 | ||
| GenBank mRNA | AK087669.1 | - | 8 | 1048 | ||
| CO795020.1 | - | 2 | 68 | |||
| DQ067445.1 | - | 8 | 1015 | |||