siRNA Details
| Gene Symbol: Fbxw15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199036.2 | NP_950201.2 | 10 | 1368 | ||
| GenBank mRNA | AK087669.1 | - | 10 | 1369 | ||
| BC099378.1 | - | 7 | 807 | 30936553 | ||
| CO795020.1 | - | 4 | 389 | |||
| DQ067445.1 | - | 10 | 1336 | |||

Fbxw15
Genomic Map