siRNA Details
| Gene Symbol: Znrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199143.2 | NP_954594.1 | 2, 3 | 653 | ||
| GenBank mRNA | AF513708.1 | - | 2, 3 | 653 | ||
| BC104316.2 | - | 2, 3 | 298 | |||
| BC104317.2 | - | 2, 3 | 307 | |||
| BC125242.1 | - | 2, 3 | 307 | |||

Znrf2
Genomic Map