siRNA Details
| Gene Symbol: Tas2r130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199156.1 | NP_954607.1 | 1 | 497 | ||
| GenBank mRNA | BC119329.1 | - | 1 | 579 | 8734078 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r130| Gene Symbol: Tas2r130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199156.1 | NP_954607.1 | 1 | 497 | ||
| GenBank mRNA | BC119329.1 | - | 1 | 579 | 8734078 | |