siRNA Details
| Gene Symbol: ALYREF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005782.3 | NP_005773.3 | 2, 3 | 404 | ||
| GenBank mRNA | AF047002.1 | - | 2, 3 | 305 | ||
| AF086513.1 | - | 1, 2 | 59 | |||
| BC052302.1 | - | 2, 3 | 404 | 6381528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALYREF| Gene Symbol: ALYREF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005782.3 | NP_005773.3 | 2, 3 | 404 | ||
| GenBank mRNA | AF047002.1 | - | 2, 3 | 305 | ||
| AF086513.1 | - | 1, 2 | 59 | |||
| BC052302.1 | - | 2, 3 | 404 | 6381528 | ||