siRNA Details
| Gene Symbol: Zhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199449.2 | NP_955520.1 | 3 | 770 | ||
| GenBank mRNA | AB099526.1 | - | 1 | 153 | ||
| AK031782.1 | - | 3 | 667 | |||
| AK157631.1 | - | 3 | 757 | |||
| BC059178.1 | - | 3 | 524 | 6400319 | ||

Zhx2
Genomic Map