siRNA Details
| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168655.1 | NP_001162126.1 | 3 | 845 | ||
| NM_001168656.1 | NP_001162127.1 | 3 | 803 | |||
| NM_001168657.1 | NP_001162128.1 | 2 | 724 | |||
| NM_007706.4 | NP_031732.1 | 3 | 872 | |||
| GenBank mRNA | AK033206.1 | - | 1 | 272 | ||
| AK137403.1 | - | 3 | 752 | |||
| AK144810.1 | - | 3 | 814 | |||
| AK160537.1 | - | 3 | 829 | |||
| AK162591.1 | - | 3 | 837 | |||
| BC106153.1 | - | 3 | 843 | 4989433 | ||
| U88327.1 | - | 2 | 632 | |||

Socs2
Genomic Map