siRNA Details
| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168655.1 | NP_001162126.1 | 3 | 890 | ||
| NM_001168656.1 | NP_001162127.1 | 3 | 848 | |||
| NM_001168657.1 | NP_001162128.1 | 2 | 769 | |||
| NM_007706.4 | NP_031732.1 | 3 | 917 | |||
| GenBank mRNA | AK033206.1 | - | 1 | 317 | ||
| AK137403.1 | - | 3 | 797 | |||
| AK144810.1 | - | 3 | 859 | |||
| AK160537.1 | - | 3 | 874 | |||
| AK162591.1 | - | 3 | 882 | |||
| BC106153.1 | - | 3 | 888 | 4989433 | ||
| U88327.1 | - | 2 | 677 | |||

Socs2
Genomic Map