siRNA Details
| Gene Symbol: Abcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013854.1 | NP_038882.1 | 17 | 1930 | ||
| GenBank mRNA | AK145549.1 | - | 8 | 1082 | ||
| AK153057.1 | - | 2 | 146 | |||
| AK183892.1 | - | 1 | 31 | |||
| BC040783.1 | - | 6 | 782 | |||
| BC046965.1 | - | 17 | 1901 | 6485365 | ||
| BC063094.1 | - | 17 | 1930 | 6516514 | ||

Abcf1
Genomic Map